missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62059MD

 View more versions of this product

Catalog No. AAH62059MD



Specifications

Specifications

4-Fluoro-2-nitrobenzeneboronic acid pinacol ester
1288978-82-4
1,3,2-dioxaborolane, 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)[N+](=O)[O-]
267.063
267.06
96%
250mg
C12H15BFNO4
UEMDKWBOUAQJJU-UHFFFAOYSA-N
2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
71744276
Solid

Chemical Identifiers

1288978-82-4
267.063
1,3,2-dioxaborolane, 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl
2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H15BFNO4
UEMDKWBOUAQJJU-UHFFFAOYSA-N
71744276
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)[N+](=O)[O-]
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

Documents