missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-2-nitroaniline 98.0+%, TCI America™
Supplier: TCI America F038025G
Specifications
4-Fluoro-2-nitroaniline | |
94°C | |
MFCD00007830 | |
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
NC1=CC=C(F)C=C1[N+]([O-])=O | |
156.12 | |
156.12 | |
Crystalline Powder |
364-78-3 | |
C6H5FN2O2 | |
25 g | |
PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
4-fluoro-2-nitroaniline | |
67769 | |
≥98.0% (GC) |
Chemical Identifiers
364-78-3 | |
156.12 | |
PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
67769 | |
NC1=CC=C(F)C=C1[N+]([O-])=O |
C6H5FN2O2 | |
MFCD00007830 | |
2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
4-fluoro-2-nitroaniline |
Safety and Handling
TSCA : No