missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Fluoro-2'-nitroacetanilide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249660050

 View more versions of this product

Catalog No. AC249660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4'-Fluoro-2'-nitroacetanilide
Glass bottle
70°C to 73°C
99+%
99.0
99% min. (GC)
FC6H3NO2NHC(O)CH3
4'-fluoro-2'-nitroacetanilide, 4-fluoro-2-nitroacetanilide, 4/'-fluoro-2/'-nitroacetanilide, acetamide,n-4-fluoro-2-nitrophenyl, labotest-bb lt00455473, labotest-bb lt03332301, maybridge1_003501, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide, n-4-fluoro-2-nitrophenyl acetamide #
UZBZEUCQENVPQB-UHFFFAOYSA-N
N-(4-fluoro-2-nitrophenyl)acetamide
136289
Crystalline Powder
Authentic
Ochre to Yellow
5g
448-39-5
100.0
C8H7FN2O3
MFCD00031517
Solubility in water: soluble in hot water. Other solubilities: insoluble in non-polar solvents,soluble in polar,solvents
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
198.16
198.16
99+%

Chemical Identifiers

448-39-5
198.16
UZBZEUCQENVPQB-UHFFFAOYSA-N
136289
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
C8H7FN2O3
MFCD00031517
4'-fluoro-2'-nitroacetanilide, 4-fluoro-2-nitroacetanilide, 4/'-fluoro-2/'-nitroacetanilide, acetamide,n-4-fluoro-2-nitrophenyl, labotest-bb lt00455473, labotest-bb lt03332301, maybridge1_003501, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide, n-4-fluoro-2-nitrophenyl acetamide #
N-(4-fluoro-2-nitrophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents