missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Fluoro-2'-nitroacetanilide, 99+%, ACROS Organics™

Chemical Identifiers

CAS 448-39-5
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.16
MDL Number MFCD00031517
InChI Key UZBZEUCQENVPQB-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-nitroacetanilide, 4-fluoro-2-nitroacetanilide, 4/'-fluoro-2/'-nitroacetanilide, acetamide,n-4-fluoro-2-nitrophenyl, labotest-bb lt00455473, labotest-bb lt03332301, maybridge1_003501, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide, n-4-fluoro-2-nitrophenyl acetamide #
PubChem CID 136289
IUPAC Name N-(4-fluoro-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC249660050
SDS View Documents
Thermo Fisher Scientific
AC249660050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC249660250
SDS View Documents
Thermo Fisher Scientific
AC249660250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

448-39-5
198.16
UZBZEUCQENVPQB-UHFFFAOYSA-N
136289
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
C8H7FN2O3
MFCD00031517
4'-fluoro-2'-nitroacetanilide, 4-fluoro-2-nitroacetanilide, 4/'-fluoro-2/'-nitroacetanilide, acetamide,n-4-fluoro-2-nitrophenyl, labotest-bb lt00455473, labotest-bb lt03332301, maybridge1_003501, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide, n-4-fluoro-2-nitrophenyl acetamide #
N-(4-fluoro-2-nitrophenyl)acetamide

Specifications

Authentic
Ochre to Yellow
5g
99.0
99% min. (GC)
FC6H3NO2NHC(O)CH3
4'-fluoro-2'-nitroacetanilide, 4-fluoro-2-nitroacetanilide, 4/'-fluoro-2/'-nitroacetanilide, acetamide,n-4-fluoro-2-nitrophenyl, labotest-bb lt00455473, labotest-bb lt03332301, maybridge1_003501, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide, n-4-fluoro-2-nitrophenyl acetamide #
UZBZEUCQENVPQB-UHFFFAOYSA-N
N-(4-fluoro-2-nitrophenyl)acetamide
136289
Crystalline Powder
4'-Fluoro-2'-nitroacetanilide, 99+%
Glass bottle
70°C to 73°C
448-39-5
100.0
C8H7FN2O3
MFCD00031517
Solubility in water: soluble in hot water. Other solubilities: insoluble in non-polar solvents,soluble in polar,solvents
CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]
198.16
198.16
99+%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS