missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%

$48.27 - $522.69

Chemical Identifiers

CAS 179062-05-6
Molecular Formula C7H4F4N2O2
Molecular Weight (g/mol) 224.115
MDL Number MFCD03094267
InChI Key WOSGMZUIHAFPSS-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitro-5-aminobenzotrifluoride, 3-amino-6-fluoro-4-nitrobenzotrifluoride, 4-fluoro-2-nitro-5-trifluoromethyl aniline, 4-fluoro-2-nitro-5-trifluoromethyl benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl phenylamine, 5-amino-2-fluoro-4-nitrobenzotrifluoride, benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl, ksc106o9t, pubchem4933
PubChem CID 2737686
IUPAC Name 4-fluoro-2-nitro-5-(trifluoromethyl)aniline
SMILES C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64928MD View Documents Alfa Aesar™
H64928MD
250mg Each for $48.27
 
AAH6492803 View Documents Alfa Aesar™
H6492803
1g Each for $127.34
 
AAH6492806 View Documents Alfa Aesar™
H6492806
5g Each for $522.69
 
Specifications

Chemical Identifiers

179062-05-6
224.115
WOSGMZUIHAFPSS-UHFFFAOYSA-N
2737686
C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F
C7H4F4N2O2
MFCD03094267
2-fluoro-4-nitro-5-aminobenzotrifluoride, 3-amino-6-fluoro-4-nitrobenzotrifluoride, 4-fluoro-2-nitro-5-trifluoromethyl aniline, 4-fluoro-2-nitro-5-trifluoromethyl benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl phenylamine, 5-amino-2-fluoro-4-nitrobenzotrifluoride, benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl, ksc106o9t, pubchem4933
4-fluoro-2-nitro-5-(trifluoromethyl)aniline

Specifications

White
179062-05-6
MFCD03094267
14360199
WOSGMZUIHAFPSS-UHFFFAOYSA-N
4-fluoro-2-nitro-5-(trifluoromethyl)aniline
2737686
Solid
4-Fluoro-2-nitro-5-(trifluoromethyl)aniline
250mg
C7H4F4N2O2
UN2811
2-fluoro-4-nitro-5-aminobenzotrifluoride, 3-amino-6-fluoro-4-nitrobenzotrifluoride, 4-fluoro-2-nitro-5-trifluoromethyl aniline, 4-fluoro-2-nitro-5-trifluoromethyl benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl phenylamine, 5-amino-2-fluoro-4-nitrobenzotrifluoride, benzenamine, 4-fluoro-2-nitro-5-trifluoromethyl, ksc106o9t, pubchem4933
C1=C(C(=CC(=C1N)[N+](=O)[O-])F)C(F)(F)F
224.115
224.11
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures