Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-n-propoxybenzeneboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-n-propoxybenzeneboronic acid
CAS 480438-60-6
Formula Weight 198
MDL Number MFCD06411288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5526903

 
alfa aesar™
H5526903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198
Chemical Name or Material 4-Fluoro-2-n-propoxybenzeneboronic acid
Physical Form Solid
Assay 97%
MDL Number MFCD06411288
CAS 480438-60-6
Quantity 1g