Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylpyridine, 96%

$81.51 - $81.51

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylpyridine
CAS 766-16-5
Formula Weight 111.12
MDL Number MFCD04114145
Beilstein 107087
Synonym 4-Fluoro-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6428403

  SDS
alfa aesar™
H6428403
1g Each for $81.51

AAH6428406

  SDS
alfa aesar™
H6428406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64284MD

  SDS
alfa aesar™
H64284MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 128° to 132°C
Formula Weight 111.12
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methylpyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Refractive Index 1.4715
Synonym 4-Fluoro-2-picoline
Storage Note 1 -30° to -10°C
Beilstein 107087
Health Hazard 1 Warning
Assay 96%
MDL Number MFCD04114145
CAS 766-16-5
Color Colorless to pale yellow
Quantity 1g

Applications
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C