missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylpyridine, 96%

Manufacturer:  Alfa Aesar™ H64284MD

 View more versions of this product

Catalog No. AAH64284MD



Description

Description

Applications
It is used in the correlations with and limitations of the α scale of solvent hydrogen bond donor acidities.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store
Specifications

Specifications

4-Fluoro-2-methylpyridine
Air Sensitive
Colorless to Yellow
766-16-5
MFCD04114145
4-fluoro-2-picoline, 4-fluoro-2-methyl-pyridine, pubchem21038, pyridine,4-fluoro-2-methyl, 4-fluoranyl-2-methyl-pyridine
CC1=NC=CC(=C1)F
111.119
111.12
96%
1.4715
128°C to 132°C
250mg
C6H6FN
107087
LEJYGRVERQVBLB-UHFFFAOYSA-N
4-fluoro-2-methylpyridine
2762819
Liquid

Chemical Identifiers

766-16-5
111.119
LEJYGRVERQVBLB-UHFFFAOYSA-N
2762819
CC1=NC=CC(=C1)F
C6H6FN
MFCD04114145
4-fluoro-2-picoline, 4-fluoro-2-methyl-pyridine, pubchem21038, pyridine,4-fluoro-2-methyl, 4-fluoranyl-2-methyl-pyridine
4-fluoro-2-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents