Please login to your online account to display your discounted pricing

4-Fluoro-2-methylphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
CAS 139911-29-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8BFO2
Formula Weight 153.95
MDL Number MFCD02093072
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367940250

  SDS
thermo fisher scientific
AC367940250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367940010

  SDS
thermo fisher scientific
AC367940010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367940050

  SDS
thermo fisher scientific
AC367940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 153.95
Quantity 25g
Color White to off-white
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
CAS 139911-29-8
MDL Number MFCD02093072
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H8BFO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.