missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylphenylacetic acid, 97%

$89.15 - $219.91

Chemical Identifiers

CAS 407640-40-8
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD00049318
Chemical Name or Material 4-Fluoro-2-methylphenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2636503 SDS Alfa Aesar™
H2636503
1g Each for $219.91
AAH26365MD SDS Alfa Aesar™
H26365MD
250mg Each for $89.15
Specifications

Description

Applications
4-Fluoro-2-methylphenylacetic acid is used as a synthetic building block.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

407640-40-8
168.17
4-Fluoro-2-methylphenylacetic acid
C9H9FO2
MFCD00049318

Specifications

96° to 99°C
407640-40-8
168.17
4-Fluoro-2-methylphenylacetic acid
1g
C9H9FO2
MFCD00049318
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning