Please login to your online account to display your discounted pricing

4-Fluoro-2-methylphenol, 98%, ACROS Organics™

$166.51 - $166.51

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylphenol
Name Note 98%
CAS 452-72-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7FO
Linear Formula FC6H3(CH3)OH
Formula Weight 126.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294370100

  SDS
thermo fisher scientific
294370100
Glass bottle 10g Each for $166.51

AC294370010

  SDS
thermo fisher scientific
AC294370010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Low Melting Solid or Liquid
Formula Weight 126.13
Color Off-white to brownish
Quantity 10g
CAS Max % 100.0
Chemical Name or Material 4-Fluoro-2-methylphenol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H7FO
Linear Formula FC6H3(CH3)OH
MDL Number MFCD00075088
CAS 452-72-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.