Please login to your online account to display your discounted pricing

4-Fluoro-2-methylbenzoic acid, 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC399300050

 View more versions of this product

Catalog No. AC399300050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.14
Quantity 5g
Chemical Name or Material 4-Fluoro-2-methylbenzoic acid
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
MDL Number MFCD04115880
CAS 321-21-1

Specifications

Chemical Name or Material 4-Fluoro-2-methylbenzoic acid
Assay Percent Range 99%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)COOH
Formula Weight 154.14
MDL Number MFCD04115880