Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid pinacol ester
CAS 815631-56-2
Formula Weight 236.09
MDL Number MFCD05863911
Synonym 4-Fluoro-2-methylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6212003

 
alfa aesar™
H6212003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6212006

 
alfa aesar™
H6212006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.09
Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid pinacol ester
Synonym 4-Fluoro-2-methylphenylboronic acid pinacol ester
Physical Form Solid
MDL Number MFCD05863911
Quantity 1g
Assay 96%
CAS 815631-56-2