missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylbenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6212006

 View more versions of this product

Catalog No. AAH6212006



Specifications

Specifications

4-Fluoro-2-methylbenzeneboronic acid pinacol ester
815631-56-2
MFCD05863911
HHWOQSZBVGPYMH-UHFFFAOYSA-N
2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2759037
Solid
5g
C13H18BFO2
1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C
236.093
236.09
96%

Chemical Identifiers

815631-56-2
236.093
HHWOQSZBVGPYMH-UHFFFAOYSA-N
2759037
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C
C13H18BFO2
MFCD05863911
1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl
2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents