Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methylanisole, 98%

$55.73 - $217.04

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylanisole
CAS 399-54-2
Molecular Formula C8H9FO
Formula Weight 140.16
MDL Number MFCD00143319
Beilstein 2517140
Synonym 5-Fluoro-2-methoxytoluene; 2-Methoxy-5-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1603106

 
alfa aesar™
A1603106
5g Each for $55.73

AAA1603114

 
alfa aesar™
A1603114
25g Each for $217.04
Description & Specifications

Specifications

Formula Weight 140.16
Chemical Name or Material 4-Fluoro-2-methylanisole
Assay 98%
Molecular Formula C8H9FO
MDL Number MFCD00143319
CAS 399-54-2
Beilstein 2517140
Synonym 5-Fluoro-2-methoxytoluene; 2-Methoxy-5-fluorotoluene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 69° to 70°C (4mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.