missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-methylaniline, TCI America™

Manufacturer:  TCI America F039825G

 View more versions of this product

Catalog No. F039825G



Specifications

Specifications

4-Fluoro-2-methylaniline
Yellow
452-71-1
MFCD00007832
KMHLGVTVACLEJE-UHFFFAOYSA-N
4-fluoro-2-methylaniline
67982
Clear Liquid at 20°C
102°C
25g
C7H8FN
2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene, 2-amino-5-fluorotoluene
CC1=C(C=CC(=C1)F)N
125.146
125.15
≥98.0% (GC,T)

Chemical Identifiers

452-71-1
125.146
KMHLGVTVACLEJE-UHFFFAOYSA-N
67982
CC1=C(C=CC(=C1)F)N
C7H8FN
MFCD00007832
2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene, 2-amino-5-fluorotoluene
4-fluoro-2-methylaniline
Safety and Handling

Safety and Handling

RTECSNumber : CY0500000

TSCA : Yes

Documents