Please login to your online account to display your discounted pricing

4-Fluoro-2-methylaniline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methylaniline
Name Note 97%
CAS 452-71-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)nH
Formula Weight 125.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369840250

  SDS
thermo fisher scientific
AC369840250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 125.15
Quantity 25g
Color Clear yellow or pink to dark red
Density 1.126g/mL
Beilstein 12, 1786
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Fluoro-2-methylaniline
Storage Information Product darkens on storage
Refractive Index 1.5350 to 1.5380 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)nH
MDL Number MFCD00007832
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Name Note 97%
CAS 452-71-1
Synonym 4-Fluoro-o-toluidine
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.