missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-methoxyphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 179899-07-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H8BFO3
Formula Weight 169.95
MDL Number MFCD03788423
Chemical Name or Material 4-Fluoro-2-methoxyphenylboronic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC434080010 SDS Thermo Fisher Scientific
AC434080010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434080050 SDS Thermo Fisher Scientific
AC434080050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

179899-07-1
100.0
C7H8BFO3
MFCD03788423
≥96.0%
96.0
97%
169.95
4-Fluoro-2-methoxyphenylboronic acid

Specifications

Crystalline Powder
1g
96.0
97%
MFCD03788423
Glass bottle
4-Fluoro-2-methoxyphenylboronic acid
169.95
White
100.0
C7H8BFO3
179899-07-1
Authentic
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.