Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
CAS 1293284-58-8
Formula Weight 280.1
Synonym 4-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6249203

 
alfa aesar™
H6249203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62492MD

 
alfa aesar™
H62492MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.1
Chemical Name or Material 4-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
CAS 1293284-58-8
Synonym 4-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester
Quantity 1g