Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methoxybenzylamine hydrochloride, 97%

$118.53 - $363.27

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methoxybenzylamine hydrochloride
CAS 870562-90-6
Molecular Formula C8H10FNO·HCl
Formula Weight 191.63
MDL Number MFCD04116342
Beilstein 11432979
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5115303

 
alfa aesar™
H5115303
1g Each for $118.53

AAH5115306

 
alfa aesar™
H5115306
5g Each for $363.27
Description & Specifications

Specifications

Formula Weight 191.63
Chemical Name or Material 4-Fluoro-2-methoxybenzylamine hydrochloride
Assay 97%
Molecular Formula C8H10FNO·HCl
MDL Number MFCD04116342
CAS 870562-90-6
Beilstein 11432979
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.