Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-methoxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-methoxybenzeneboronic acid pinacol ester
CAS 624741-83-9
Formula Weight 252.09
Synonym 4-Fluoro-2-methoxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6239103

  SDS
alfa aesar™
H6239103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62391MD

  SDS
alfa aesar™
H62391MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.09
CAS 624741-83-9
Chemical Name or Material 4-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Physical Form Solid
Quantity 1g
Synonym 4-Fluoro-2-methoxyphenylboronic acid pinacol ester
Assay 96%