missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-isopropoxybenzeneboronic acid, 97%

$226.85 - $680.54

Chemical Identifiers

CAS 480438-59-3
Molecular Formula C9H12BFO3
Molecular Weight (g/mol) 198
MDL Number MFCD06411287
InChI Key RSEFJQODCQCGRB-UHFFFAOYSA-N
Synonym 4-fluoro-2-isopropoxybenzene boronic acid
PubChem CID 16217202
IUPAC Name (4-fluoro-2-propan-2-yloxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55338MD View Documents Alfa Aesar™
H55338MD
250mg Each for $226.85
 
AAH5533803 View Documents Alfa Aesar™
H5533803
1g Each for $680.54
 
Specifications

Chemical Identifiers

480438-59-3
198
RSEFJQODCQCGRB-UHFFFAOYSA-N
16217202
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
C9H12BFO3
MFCD06411287
4-fluoro-2-isopropoxybenzene boronic acid
(4-fluoro-2-propan-2-yloxyphenyl)boronic acid

Specifications

250mg
C9H12BFO3
4-fluoro-2-isopropoxybenzene boronic acid
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
198
198
97%
480438-59-3
MFCD06411287
RSEFJQODCQCGRB-UHFFFAOYSA-N
(4-fluoro-2-propan-2-yloxyphenyl)boronic acid
16217202
Solid
4-Fluoro-2-isopropoxybenzeneboronic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures