Alfa Aesar™ 4-Fluoro-2-isopropoxybenzeneboronic acid, 97%
Manufacturer:  Alfa Aesar™ H55338MD
 View more versions of this product
Catalog No. AAH55338MD
Specifications
Specifications
4-Fluoro-2-isopropoxybenzeneboronic acid
480438-59-3
MFCD06411287
RSEFJQODCQCGRB-UHFFFAOYSA-N
(4-fluoro-2-propan-2-yloxyphenyl)boronic acid
16217202
Solid
250mg
C9H12BFO3
4-fluoro-2-isopropoxybenzene boronic acid
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
198
198
97%
Chemical Identifiers
480438-59-3
198
RSEFJQODCQCGRB-UHFFFAOYSA-N
16217202
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
C9H12BFO3
MFCD06411287
4-fluoro-2-isopropoxybenzene boronic acid
(4-fluoro-2-propan-2-yloxyphenyl)boronic acid
Safety and Handling
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
Documents