Alfa Aesar™ 4-Fluoro-2-iodotoluene, 98%
Manufacturer:  Alfa Aesar™ L0683306
 View more versions of this product
Catalog No. AAL0683306
Specifications
Specifications
4-Fluoro-2-iodotoluene
206°C to 208°C
13194-67-7
MFCD00013709
4-fluoro-2-iodotoluene, benzene, 4-fluoro-2-iodo-1-methyl, 2-iodo-4-fluorotoluene, 4-fluoro-2-iodo-1-methyl-benzene, 1-fluoro-3-iodo-4-methyl-benzene, benzene, 4-fluoro-2-iodo-1-methyl, 4-fluor-2-iod-1-methylbenzol, zlchem 432, pubchem1622, acmc-209bo9
CC1=C(C=C(C=C1)F)I
236.028
236.03
87°C (189°F)
5g
C7H6FI
2242594
RZGYAMQMAVTAKP-UHFFFAOYSA-N
4-fluoro-2-iodo-1-methylbenzene
83220
98%
Chemical Identifiers
13194-67-7
236.028
RZGYAMQMAVTAKP-UHFFFAOYSA-N
83220
CC1=C(C=C(C=C1)F)I
C7H6FI
MFCD00013709
4-fluoro-2-iodotoluene, benzene, 4-fluoro-2-iodo-1-methyl, 2-iodo-4-fluorotoluene, 4-fluoro-2-iodo-1-methyl-benzene, 1-fluoro-3-iodo-4-methyl-benzene, benzene, 4-fluoro-2-iodo-1-methyl, 4-fluor-2-iod-1-methylbenzol, zlchem 432, pubchem1622, acmc-209bo9
4-fluoro-2-iodo-1-methylbenzene
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
Documents