Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-iodotoluene, 98%

$91.27 - $344.80

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-iodotoluene
CAS 13194-67-7
Molecular Formula C7H6FI
Formula Weight 236.03
MDL Number MFCD00013709
Beilstein 2242594
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0683306

 
alfa aesar™
L0683306
5g Each for $91.27

AAL0683314

 
alfa aesar™
L0683314
25g Each for $344.80
Description & Specifications

Specifications

Formula Weight 236.03
Chemical Name or Material 4-Fluoro-2-iodotoluene
Molecular Formula C7H6FI
MDL Number MFCD00013709
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Beilstein 2242594
Flash Point 87°C (189°F)
Boiling Point 206° to 208°C
CAS 13194-67-7
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.