Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-iodotoluene, 98%


Quantity 25g

Manufacturer: alfa aesar™  L0683314

 View more versions of this product

Catalog No. AAL0683314

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 236.03
Chemical Name or Material 4-Fluoro-2-iodotoluene
Assay 98%
MDL Number MFCD00013709
CAS 13194-67-7
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Beilstein 2242594
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Molecular Formula C7H6FI
Flash Point 87°C (189°F)
Boiling Point 206° to 208°C

Specifications

Chemical Name or Material 4-Fluoro-2-iodotoluene
CAS 13194-67-7
Molecular Formula C7H6FI
Formula Weight 236.03
MDL Number MFCD00013709
Beilstein 2242594
Accessories

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.