missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-iodobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H5553103

 View more versions of this product

Catalog No. AAH5553103



Specifications

Specifications

4-Fluoro-2-iodobenzoic acid
143°C to 147°C
56096-89-0
MFCD04108278
2-iodo-4-fluorobenzoic acid
C1=CC(=C(C=C1F)I)C(=O)O
266.01
266.01
Light sensitive
1g
C7H4FIO2
UN2811
DUKFTVLJAXWGPI-UHFFFAOYSA-N
4-fluoro-2-iodobenzoic acid
12520164
97%

Chemical Identifiers

56096-89-0
266.01
DUKFTVLJAXWGPI-UHFFFAOYSA-N
12520164
C1=CC(=C(C=C1F)I)C(=O)O
C7H4FIO2
MFCD04108278
2-iodo-4-fluorobenzoic acid
4-fluoro-2-iodobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents