missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H62252MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester | |
C12H16BFO3 | |
250mg | |
PSQUBOBJDBVJIN-UHFFFAOYSA-N | |
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
72219414 | |
96% |
1038828-32-8 | |
MFCD16994248 | |
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1 | |
238.07 | |
238.06 |
Chemical Identifiers
1038828-32-8 | |
238.07 | |
PSQUBOBJDBVJIN-UHFFFAOYSA-N | |
72219414 | |
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1 |
C12H16BFO3 | |
MFCD16994248 | |
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures