Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-2-formylbenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-formylbenzeneboronic acid pinacol ester
Formula Weight 250.07
Synonym 4-Fluoro-2-formylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6210403

 
alfa aesar™
H6210403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62104MD

 
alfa aesar™
H62104MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.07
Chemical Name or Material 4-Fluoro-2-formylbenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Synonym 4-Fluoro-2-formylphenylboronic acid pinacol ester
Quantity 1g