Please login to your online account to display your discounted pricing

4-Fluoro-2-(1H-pyrazol-3-yl)phenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
CAS 288401-64-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7FN2O
Formula Weight 178.17
Synonym 3-(5-Fluoro-2-hydroxyphenyl)pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439060050

 
thermo fisher scientific
AC439060050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439060010

 
thermo fisher scientific
AC439060010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥96.0%
Formula Weight 178.17
Quantity 5g
Color White to yellow
Melting Point 130° to 135°C
CAS Min % 96.0
CAS Max % 100.0
Synonym 3-(5-Fluoro-2-hydroxyphenyl)pyrazole
Chemical Name or Material 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C9H7FN2O
CAS 288401-64-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.