missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-(1H-pyrazol-3-yl)phenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439060050

 View more versions of this product

Catalog No. AC439060050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluoro-2-(1H-pyrazol-3-yl)phenol
White to Yellow
5g
96.0
96% min. (HPLC)
3-5-fluoro-2-hydroxyphenyl pyrazole, 4-fluoro-2-1h-pyrazol-3-yl phenol, 6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one, pyrazole, 31
C1=CC(=O)C(=C2C=CNN2)C=C1F
178.17
178.17
97%
Authentic
132.9°C to 134.1°C
288401-64-9
100.0
C9H7FN2O
DIVXDXZKHCYBPY-FPLPWBNLSA-N
(6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
16217010
Powder or Crystalline Powder

Chemical Identifiers

288401-64-9
178.17
3-5-fluoro-2-hydroxyphenyl pyrazole, 4-fluoro-2-1h-pyrazol-3-yl phenol, 6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one, pyrazole, 31
(6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
C9H7FN2O
DIVXDXZKHCYBPY-FPLPWBNLSA-N
16217010
C1=CC(=O)C(=C2C=CNN2)C=C1F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents