Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-1-iodo-2-nitrobenzene, 97%

$177.80 - $177.80

Chemical Identifiers

Chemical Name or Material 4-Fluoro-1-iodo-2-nitrobenzene
CAS 364-77-2
Molecular Formula C6H3FINO2
Formula Weight 267
MDL Number MFCD00153169
Beilstein 3253380
Synonym 5-Fluoro-2-iodonitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1896206

  SDS
alfa aesar™
A1896206
5g Each for $177.80
Description & Specifications

Specifications

Formula Weight 267
Synonym 5-Fluoro-2-iodonitrobenzene
Health Hazard 1 Danger
Boiling Point 110°C (4mmHg)
MDL Number MFCD00153169
Beilstein 3253380
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Molecular Formula C6H3FINO2
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Chemical Name or Material 4-Fluoro-1-iodo-2-nitrobenzene
CAS 364-77-2
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.