Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-1,3-phenylenediamine, 95%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-1,3-phenylenediamine
CAS 6264-67-1
Formula Weight 126.13
MDL Number MFCD00077494
Synonym 4-Fluoro-1,3-diaminobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6147603

  SDS
alfa aesar™
H6147603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6147606

  SDS
alfa aesar™
H6147606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 152° to 153°C
Formula Weight 126.13
Melting Point 34° to 37°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Color Black
Quantity 1g
Chemical Name or Material 4-Fluoro-1,3-phenylenediamine
MDL Number MFCD00077494
Physical Form Powder
CAS 6264-67-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4-Fluoro-1,3-diaminobenzene
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.