missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethynyltoluene, TCI America™

Manufacturer:  TCI America E065525G

 View more versions of this product

Catalog No. E065525G



Specifications

Specifications

4-Ethynyltoluene
Undesignated
766-97-2
1325
KSZVOXHGCKKOLL-UHFFFAOYSA-N
1-ethynyl-4-methylbenzene
13018
Crystal-Lump at 20°C
168°C
25g
C9H8
4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
CC1=CC=C(C=C1)C#C
116.163
116.16
≥98.0% (GC)

Chemical Identifiers

766-97-2
116.163
4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
1-ethynyl-4-methylbenzene
C9H8
KSZVOXHGCKKOLL-UHFFFAOYSA-N
13018
CC1=CC=C(C=C1)C#C
Safety and Handling

Safety and Handling

RTECSNumber : XT2570000

TSCA : No

Documents