missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethynylphenylacetonitrile, 95%

$965.47 - $965.47

Chemical Identifiers

CAS 351002-90-9
Molecular Formula C10H7N
Molecular Weight (g/mol) 141.173
MDL Number MFCD05664204
InChI Key CZUBOLAETAAHLA-UHFFFAOYSA-N
Synonym 2-4-ethynylphenyl acetonitrile, 4-ethynylbenzeneacetonitrile, 4-ethynylphenyl acetonitrile, 4-ethynylphenylacetonitrile, benzeneacetonitrile, 4-ethynyl, ksc497s1l
PubChem CID 16069314
IUPAC Name 2-(4-ethynylphenyl)acetonitrile
SMILES C#CC1=CC=C(C=C1)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6112214 View Documents Alfa Aesar™
H6112214
25g Each for $965.47
 
Description

Description

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Chemical Identifiers

351002-90-9
141.173
CZUBOLAETAAHLA-UHFFFAOYSA-N
16069314
C#CC1=CC=C(C=C1)CC#N
C10H7N
MFCD05664204
2-4-ethynylphenyl acetonitrile, 4-ethynylbenzeneacetonitrile, 4-ethynylphenyl acetonitrile, 4-ethynylphenylacetonitrile, benzeneacetonitrile, 4-ethynyl, ksc497s1l
2-(4-ethynylphenyl)acetonitrile

Specifications

1.037 g/mL
1.563
351002-90-9
MFCD05664204
2-4-ethynylphenyl acetonitrile, 4-ethynylbenzeneacetonitrile, 4-ethynylphenyl acetonitrile, 4-ethynylphenylacetonitrile, benzeneacetonitrile, 4-ethynyl, ksc497s1l
CZUBOLAETAAHLA-UHFFFAOYSA-N
2-(4-ethynylphenyl)acetonitrile
16069314
Liquid
4-Ethynylphenylacetonitrile
>110°C (230°F)
25g
C10H7N
UN3276
Slightly soluble in water.
C#CC1=CC=C(C=C1)CC#N
141.173
141.17
95%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C