missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethynylphenylacetonitrile, 95%

Manufacturer:  Alfa Aesar™ H6112214

 View more versions of this product

Catalog No. AAH6112214



Description

Description

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Specifications

4-Ethynylphenylacetonitrile
>110°C (230°F)
25g
C10H7N
UN3276
Slightly soluble in water.
C#CC1=CC=C(C=C1)CC#N
141.173
141.17
95%
1.037 g/mL
1.563
351002-90-9
MFCD05664204
2-4-ethynylphenyl acetonitrile, 4-ethynylbenzeneacetonitrile, 4-ethynylphenyl acetonitrile, 4-ethynylphenylacetonitrile, benzeneacetonitrile, 4-ethynyl, ksc497s1l
CZUBOLAETAAHLA-UHFFFAOYSA-N
2-(4-ethynylphenyl)acetonitrile
16069314
Liquid

Chemical Identifiers

351002-90-9
141.173
CZUBOLAETAAHLA-UHFFFAOYSA-N
16069314
C#CC1=CC=C(C=C1)CC#N
C10H7N
MFCD05664204
2-4-ethynylphenyl acetonitrile, 4-ethynylbenzeneacetonitrile, 4-ethynylphenyl acetonitrile, 4-ethynylphenylacetonitrile, benzeneacetonitrile, 4-ethynyl, ksc497s1l
2-(4-ethynylphenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents