missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethynylphenylacetonitrile, 95%

Manufacturer:  Alfa Aesar™ H6112206

 View more versions of this product

Catalog No. AAH6112206



Description

Description

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Specifications

4-Ethynylphenylacetonitrile
5g
C10H7N
UN3276
CZUBOLAETAAHLA-UHFFFAOYSA-N
2-(4-ethynylphenyl)acetonitrile
16069314
Liquid
>110°C (230°F)
351002-90-9
MFCD05664204
4-ethynylphenylacetonitrile, 2-4-ethynylphenyl acetonitrile, 4-ethynylphenyl acetonitrile, benzeneacetonitrile, 4-ethynyl, 4-ethynylbenzeneacetonitrile, ksc497s1l, 4-ethynylphenylacetonitrile
C#CC1=CC=C(C=C1)CC#N
141.173
141.17
95%

Chemical Identifiers

351002-90-9
141.173
CZUBOLAETAAHLA-UHFFFAOYSA-N
16069314
C#CC1=CC=C(C=C1)CC#N
C10H7N
MFCD05664204
4-ethynylphenylacetonitrile, 2-4-ethynylphenyl acetonitrile, 4-ethynylphenyl acetonitrile, benzeneacetonitrile, 4-ethynyl, 4-ethynylbenzeneacetonitrile, ksc497s1l, 4-ethynylphenylacetonitrile
2-(4-ethynylphenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

storageNote1 : -30° to -10°C

Recommended Storage : −30° to −10°C

Documents