missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethynylbenzaldehyde, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC432940050

 View more versions of this product

Catalog No. AC432940050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Ethynylbenzaldehyde
Glass bottle
89.0°C to 94.5°C
63697-96-1
100.0
C9H6O
BGMHQBQFJYJLBP-UHFFFAOYSA-N
4-ethynylbenzaldehyde
2771645
Powder or Crystalline Powder
Authentic
Brown to White
5g
94.0
95%
4-ethynyl benzaldehyde, 4-ethynyl-benzaldehyde, benzaldehyde, 4-ethynyl, benzaldehyde,4-ethynyl, p-ethynylbenzaldehyde, 4-acetylenylbenzaldehyde, acmc-209x0m, 4-ethynylbenzaldehyde, benzaldehyde, 4-ethynyl-9ci
C#CC1=CC=C(C=C1)C=O
130.146
130.15
95%

Chemical Identifiers

63697-96-1
130.146
4-ethynyl benzaldehyde, 4-ethynyl-benzaldehyde, benzaldehyde, 4-ethynyl, benzaldehyde,4-ethynyl, p-ethynylbenzaldehyde, 4-acetylenylbenzaldehyde, acmc-209x0m, 4-ethynylbenzaldehyde, benzaldehyde, 4-ethynyl-9ci
4-ethynylbenzaldehyde
C9H6O
BGMHQBQFJYJLBP-UHFFFAOYSA-N
2771645
C#CC1=CC=C(C=C1)C=O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents