missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethynylanisole 98.0+%, TCI America™
Supplier: TCI America E060325G
Specifications
| 4-Ethynylanisole | |
| White-Yellow | |
| C9H8O | |
| 25 g | |
| KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
| 1-ethynyl-4-methoxybenzene | |
| 251020 | |
| ≥98.0% (GC) |
| 768-60-5 | |
| 86°C | |
| MFCD00168815 | |
| 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
| COC1=CC=C(C=C1)C#C | |
| 132.162 | |
| 132.16 | |
| Crystalline Lumps |
Chemical Identifiers
| 768-60-5 | |
| 132.162 | |
| KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
| 251020 | |
| COC1=CC=C(C=C1)C#C |
| C9H8O | |
| MFCD00168815 | |
| 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
| 1-ethynyl-4-methoxybenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator