Please login to your online account to display your discounted pricing

4'-Ethynylacetophenone 98% , ACROS Organics™

$767.91 - $767.91

Chemical Identifiers

Chemical Name or Material 4'-Ethynylacetophenone
CAS 42472-69-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H8O
Linear Formula CHCC6H4COCH3
Formula Weight 144.17
MDL Number MFCD04974025
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390580010

  SDS
fisher scientific acros
AC390580010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390580050

  SDS
thermo fisher scientific
AC390580050
Glass bottle 5g Each for $767.91
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 144.17
Physical Form Powder
Quantity 1g
Color Yellow
Melting Point 67° to 70°C
CAS Max % 100.0
Linear Formula CHCC6H4COCH3
Molecular Formula C10H8O
Chemical Name or Material 4'-Ethynylacetophenone
Assay Percent Range 98%
MDL Number MFCD04974025
CAS Min % 97.5
CAS 42472-69-5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.