Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethynyl-2,6-dimethyl-4-heptanol, 97%

$67.11 - $237.05

Chemical Identifiers

Chemical Name or Material 4-Ethynyl-2,6-dimethyl-4-heptanol
CAS 10562-68-2
Molecular Formula C11H20O
Formula Weight 168.28
MDL Number MFCD00041605
Beilstein 1752640
Synonym Ethynyl diisobutyl carbinol; 3-Isobutyl-5-methyl-1-hexyn-3-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1244604

 
alfa aesar™
L1244604
2g Each for $67.11

AAL1244609

 
alfa aesar™
L1244609
10g Each for $237.05
Description & Specifications

Specifications

Formula Weight 168.28
Assay 97%
Molecular Formula C11H20O
MDL Number MFCD00041605
CAS 10562-68-2
Beilstein 1752640
Synonym Ethynyl diisobutyl carbinol; 3-Isobutyl-5-methyl-1-hexyn-3-ol
Boiling Point 83° to 84°C (18mmHg)
Chemical Name or Material 4-Ethynyl-2,6-dimethyl-4-heptanol
Quantity 2g