missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Ethylpropiophenone, TCI America™

Manufacturer:  TCI America E0461500ML

 View more versions of this product

Catalog No. E0461500ML



Specifications

Specifications

4′-Ethylpropiophenone
Colorless
27465-51-6
MFCD00191646
VGQRIILEZYZAOE-UHFFFAOYSA-N
1-(4-ethylphenyl)propan-1-one
583750
Clear Liquid at 20°C
246°C
500mL
C11H14O
Ethyl 4-Ethylphenyl Ketone
CCC1=CC=C(C=C1)C(=O)CC
162.232
162.23
≥96.0% (GC)

Chemical Identifiers

27465-51-6
162.232
VGQRIILEZYZAOE-UHFFFAOYSA-N
583750
CCC1=CC=C(C=C1)C(=O)CC
C11H14O
MFCD00191646
Ethyl 4-Ethylphenyl Ketone
1-(4-ethylphenyl)propan-1-one
Safety and Handling

Safety and Handling

TSCA : No

Documents