missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylphenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 359320050

 View more versions of this product

Catalog No. AC359320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Ethylphenylboronic acid
96.0
Authentic
Glass bottle
5g
Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,methanol
B(C1=CC=C(C=C1)CC)(O)O
149.98
149.98
63139-21-9
100.0
96% min. (HPLC)
C8H11BO2
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
RZCPLOMUUCFPQA-UHFFFAOYSA-N
(4-ethylphenyl)boronic acid
2734352
97%

Chemical Identifiers

63139-21-9
149.98
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
(4-ethylphenyl)boronic acid
C8H11BO2
RZCPLOMUUCFPQA-UHFFFAOYSA-N
2734352
B(C1=CC=C(C=C1)CC)(O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only