Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylphenylacetone, 97+%

$210.88 - $210.88

Chemical Identifiers

Chemical Name or Material 4-Ethylphenylacetone
CAS 75251-24-0
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00210429
Synonym 1-(4-Ethylphenyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1881006

 
alfa aesar™
A1881006
5g Each for $210.88
Description & Specifications

Specifications

Formula Weight 162.23
Chemical Name or Material 4-Ethylphenylacetone
Assay ≥97%
Molecular Formula C11H14O
MDL Number MFCD00210429
CAS 75251-24-0
Synonym 1-(4-Ethylphenyl)-2-propanone
Quantity 5g