missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylphenyl trans-4-(4-n-pentylcyclohexyl)benzoate, 99%

Manufacturer:  Alfa Aesar™ H5641803

 View more versions of this product

Catalog No. AAH5641803



Specifications

Specifications

4-Ethylphenyl trans-4-(4-n-pentylcyclohexyl)benzoate
91223-44-8
MFCD16879074
WLPXTIALVPOMAH-UHFFFAOYSA-N
(4-ethylphenyl) 4-(4-pentylcyclohexyl)benzoate
23334599
99%
1g
C26H34O2
4-ethylphenyl 4-trans-4-pentylcyclohexyl benzoate, 4-ethylphenyl 4-4-pentylcyclohexyl benzoate, 4-ethylphenyl 4-4-pentylcyclohexyl benzoate, 4-ethylphenyl trans-4-4-n-pentylcyclohexyl benzoate, 4-ethylphenyl 4-1s,4r-4-pentylcyclohexyl benzoate
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)CC
378.556
378.55

Chemical Identifiers

91223-44-8
378.556
WLPXTIALVPOMAH-UHFFFAOYSA-N
23334599
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)CC
C26H34O2
MFCD16879074
4-ethylphenyl 4-trans-4-pentylcyclohexyl benzoate, 4-ethylphenyl 4-4-pentylcyclohexyl benzoate, 4-ethylphenyl 4-4-pentylcyclohexyl benzoate, 4-ethylphenyl trans-4-4-n-pentylcyclohexyl benzoate, 4-ethylphenyl 4-1s,4r-4-pentylcyclohexyl benzoate
(4-ethylphenyl) 4-(4-pentylcyclohexyl)benzoate
Documents