missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylphenoxyacetic acid, 97%

$60.59 - $191.22

Chemical Identifiers

CAS 24431-27-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00014366
InChI Key WVELHLIHMYYZAT-UHFFFAOYSA-N
Synonym 4-ethylphenoxy acetic acid, 2-4-ethylphenoxy acetic acid, 4-ethylphenoxyacetic acid, 2-p-ethylphenoxy acetic acid, acetic acid, 4-ethylphenoxy, acmc-1cdn7, p-ethylphenoxy acetic acid, cambridge id 5147400, acetic acid,2-4-ethylphenoxy
PubChem CID 90499
IUPAC Name 2-(4-ethylphenoxy)acetic acid
SMILES CCC1=CC=C(C=C1)OCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0110206 View Documents Alfa Aesar™
L0110206
5g Each for $60.59
 
AAL0110214 View Documents Alfa Aesar™
L0110214
25g Each for $191.22
 
Specifications

Chemical Identifiers

24431-27-4
180.203
WVELHLIHMYYZAT-UHFFFAOYSA-N
90499
CCC1=CC=C(C=C1)OCC(=O)O
C10H12O3
MFCD00014366
4-ethylphenoxy acetic acid, 2-4-ethylphenoxy acetic acid, 4-ethylphenoxyacetic acid, 2-p-ethylphenoxy acetic acid, acetic acid, 4-ethylphenoxy, acmc-1cdn7, p-ethylphenoxy acetic acid, cambridge id 5147400, acetic acid,2-4-ethylphenoxy
2-(4-ethylphenoxy)acetic acid

Specifications

92°C to 94°C
24431-27-4
MFCD00014366
4-ethylphenoxy acetic acid, 2-4-ethylphenoxy acetic acid, 4-ethylphenoxyacetic acid, 2-p-ethylphenoxy acetic acid, acetic acid, 4-ethylphenoxy, acmc-1cdn7, p-ethylphenoxy acetic acid, cambridge id 5147400, acetic acid,2-4-ethylphenoxy
CCC1=CC=C(C=C1)OCC(=O)O
180.203
180.2
4-Ethylphenoxyacetic acid
5g
C10H12O3
2259506
WVELHLIHMYYZAT-UHFFFAOYSA-N
2-(4-ethylphenoxy)acetic acid
90499
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning