missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylphenol, TCI America™

Manufacturer:  TCI America E015925G

 View more versions of this product

Catalog No. E015925G



Specifications

Specifications

4-Ethylphenol
White-Yellow
123-07-9
MFCD00002393
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
CCC1=CC=C(C=C1)O
122.167
CHEBI:49584
Crystal-Lump at 20°C
219°C
25g
C8H10O
2430
HXDOZKJGKXYMEW-UHFFFAOYSA-N
4-ethylphenol
31242
122.17
≥97.0% (GC)

Chemical Identifiers

123-07-9
122.167
HXDOZKJGKXYMEW-UHFFFAOYSA-N
31242
4-ethylphenol
C8H10O
MFCD00002393
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
CHEBI:49584
CCC1=CC=C(C=C1)O
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0500&(4)-0057

RTECSNumber : SL4040000

TSCA : Yes

Documents