missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylphenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC118820051

 View more versions of this product

Catalog No. AC118820051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Ethylphenol
Authentic
218.0°C to 219.0°C
41.0°C to 46.0°C
123-07-9
100.0
C8H10O
06,472
Solubility in water: 4900mg/l (25°C). Other solubilities: soluble in alcohol and ether
CCC1=CC=C(C=C1)O
122.17
CHEBI:49584
Crystalline Mass or Crystals
100°C
Plastic drum
Brown to White
5kg
96.0
96% min. (GC)
C2H5C6H4OH
1-ethyl-4-hydroxybenzene, 1-hydroxy-4-ethylbenzene, 4-ethyl-phenol, 4-hydroxyethylbenzene, 4-hydroxyphenylethane, p-ethylphenol, para-ethylphenol, paraethylphenol, phenol, 4-ethyl, phenol, p-ethyl
HXDOZKJGKXYMEW-UHFFFAOYSA-N
4-ethylphenol
31242
122.17
97%

Chemical Identifiers

123-07-9
122.17
1-ethyl-4-hydroxybenzene, 1-hydroxy-4-ethylbenzene, 4-ethyl-phenol, 4-hydroxyethylbenzene, 4-hydroxyphenylethane, p-ethylphenol, para-ethylphenol, paraethylphenol, phenol, 4-ethyl, phenol, p-ethyl
CHEBI:49584
CCC1=CC=C(C=C1)O
C8H10O
HXDOZKJGKXYMEW-UHFFFAOYSA-N
31242
4-ethylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-598-6

SDS Documents