Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylphenol, 97%

Chemical Identifiers

Chemical Name or Material 4-Ethylphenol
CAS 123-07-9
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002393
UN Number UN2430
Beilstein 1363317
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2343330

 
alfa aesar™
B2343330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Molecular Formula C8H10O
MDL Number MFCD00002393
Health Hazard 2 GHS H Statement
H314-H318-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
RTECS Number SL4040000
Flash Point 100°C (212°F)
Boiling Point 218° to 219°C
Melting Point 40° to 45°C
Quantity 250g
Chemical Name or Material 4-Ethylphenol
TSCA TSCA
Assay 97%
CAS 123-07-9
UN Number UN2430
Beilstein 1363317
Health Hazard 1 Danger

Applications
4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A receptor agonists endowed with antinociceptive activity in vivo. It is also used in the preparation of isopropylamides as potent and noncovalent proteosome inhibitors.

Solubility
Soluble in water. (4.9 g/L) at 25°C

Notes
Do not store together with acids. Store away from acid chlorides and acid anhydrides. Incompatible with oxidizing agents and acid chlorides.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.