Learn More
4-Ethylphenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2343330
Description
4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A receptor agonists endowed with antinociceptive activity. It is also used in the preparation of isopropylamides as potent and noncovalent proteosome inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A recep
Specifications
4-Ethylphenol | |
40°C to 45°C | |
218°C to 219°C | |
C8H10O | |
MFCD00002393 | |
UN2430 | |
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol | |
HXDOZKJGKXYMEW-UHFFFAOYSA-N | |
4-ethylphenol | |
31242 | |
122.17 |
123-07-9 | |
1.011 | |
100°C (212°F) | |
1.533 | |
250 g | |
1363317 | |
Soluble in water. (4.9g/L) at 25°C | |
CCC1=CC=C(C=C1)O | |
122.167 | |
CHEBI:49584 | |
97% |
Chemical Identifiers
123-07-9 | |
122.167 | |
HXDOZKJGKXYMEW-UHFFFAOYSA-N | |
31242 | |
4-ethylphenol |
C8H10O | |
MFCD00002393 | |
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol | |
CHEBI:49584 | |
CCC1=CC=C(C=C1)O |
Safety and Handling
GHS H Statement
H314-H318-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber : 204-598-6
RTECSNumber : SL4040000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only